CAS RN 9024-52-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Aldolase, MP Biomedicals™
CAS: 9024-52-6 Synonym: D -Fructose-1, 6-bisphosphate-D -glyceraldehyde-3-phosphate lyase,E.C. 4.1.2.13
2
Aldolase, MP Biomedicals™
CAS: 9024-52-6 Synonym: D -Fructose-1, 6-bisphosphate-D -glyceraldehyde-3-phosphate lyase,E.C. 4.1.2.13