accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrOOH

CAS RN 90259-31-7

IUPAC Name: 2-bromo-6-methylbenzoic acid
Synonyms: 2-bromo-6-methylbenzoic acid
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: ICXBPDJQFPIBSS-UHFFFAOYSA-N
SMILES: CC1=C(C(O)=O)C(Br)=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)