accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 902779-59-3

IUPAC Name: dichlorocopper; {[(4-oxo-4H-chromen-3-yl)methylidene]amino}thiourea
Synonyms: [(4-oxochromen-3-yl)methylidene]aminothiourea; copper chloride
Molecular Weight (g/mol): 381.72
Molecular Formula: C11H9Cl2CuN3O2S
InChi Key: HZRXQEVFXXQYFS-UHFFFAOYSA-L
SMILES: Cl[Cu]Cl.NC(=S)NN=CC1=COC2=CC=CC=C2C1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results