accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 902836-40-2

IUPAC Name: 4-(4-phenyl-1H-pyrazol-1-yl)piperidine
Synonyms: 4-(4-phenylpyrazol-1-yl)piperidine
Molecular Weight (g/mol): 227.31
Molecular Formula: C14H17N3
InChi Key: SVKHSGABZPOEQY-UHFFFAOYSA-N
SMILES: C1CC(CCN1)N1C=C(C=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results