accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 902836-93-5

IUPAC Name: 3-(piperidin-3-yloxy)benzonitrile
Synonyms: 3-(piperidin-3-yloxy)benzonitrile
Molecular Weight (g/mol): 202.26
Molecular Formula: C12H14N2O
InChi Key: OSURJDMYHKQISX-UHFFFAOYNA-N
SMILES: N#CC1=CC(OC2CCCNC2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results