accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3H3CCH3OHHOH3CCH3CH3H3CCH3

CAS RN 903-19-5

IUPAC Name: 2,5-bis(2,4,4-trimethylpentan-2-yl)benzene-1,4-diol
Synonyms: 2,5-bis(2,4,4-trimethylpentan-2-yl)benzene-1,4-diol
Molecular Weight (g/mol): 334.54
Molecular Formula: C22H38O2
InChi Key: CLDZVCMRASJQFO-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)CC(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)