accessibility menu, dialog, popup

UserName

Viewing profile as user:
A(a)

CAS RN 9037-24-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula: MFCD00145841
InChi Key:
SMILES:
Color 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)