CAS RN 9041-35-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Supelco™ Sephadex™ G-25, Fine
Sephadex™ G-25 is a gel filtration medium used in affinity chromatography, protein chromatography and gel filtration chromatography.
2
MilliporeSigma™ Supelco™ Sephadex™ G-25, Coarse
Sephadex™ G-25 is a gel filtration medium used in affinity chromatography, protein chromatography and gel filtration chromatography.