CAS RN 90417-38-2
IUPAC Name:
8-hydroxy-2-methyl-1,4-dihydroquinazolin-4-one
Synonyms:
8-hydroxy-2-methyl-1H-quinazolin-4-one
Molecular Weight (g/mol):
176.18
Molecular Formula:
C9H8N2O2
InChi Key:
YJDAOHJWLUNFLX-UHFFFAOYSA-N
SMILES:
CC1=NC(=O)C2=CC=CC(O)=C2N1
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PARP Inhibitor VI, NU1025
The PARP Inhibitor VI, NU1025, also referenced under CAS 90417-38-2, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.