CAS RN 9042-11-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Amberlite™ CG-50 - type 1, ion-exchange resin
CAS: 9042-11-9 MDL Number: MFCD00132693