accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOFF

CAS RN 90446-25-6

IUPAC Name: 4-(difluoromethoxy)benzonitrile
Synonyms: 4-(difluoromethoxy)benzonitrile
Molecular Weight (g/mol): 169.13
Molecular Formula: C8H5F2NO
InChi Key: OGMOYCCCAFJQKI-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results