accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHO

CAS RN 90484-22-3

IUPAC Name: 1-benzofuran-7-carboxylic acid
Synonyms: 1-benzofuran-7-carboxylic acid
Molecular Weight (g/mol): 162.14
Molecular Formula: C9H6O3
InChi Key: QMHILIQFOBNARN-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2OC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results