accessibility menu, dialog, popup

UserName

Viewing profile as user:
IH3CNOONH3C(Z)(E/Z)

CAS RN 905-96-4

IUPAC Name: 3-ethyl-2-{3-[(2Z)-3-ethyl-2,3-dihydro-1,3-benzoxazol-2-ylidene]prop-1-en-1-yl}-1,3-benzoxazol-3-ium iodide
Synonyms: 3-ethyl-2-{3-[(2Z)-3-ethyl-1,3-benzoxazol-2-ylidene]prop-1-en-1-yl}-1,3-benzoxazol-3-ium iodide
Molecular Weight (g/mol): 460.32
Molecular Formula: C21H21IN2O2
InChi Key: FIZZUEJIOKEFFZ-UHFFFAOYSA-M
SMILES: [I-].CCN1\C(OC2=CC=CC=C12)=C\C=CC1=[N+](CC)C2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)