CAS RN 9050-94-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Supelco™ Sephadex™ G-100, Superfine
Sephadex™ G-100 is a gel filtration medium used in protein chromatography, affinity chromatography and gel filtration chromatography.
2
MilliporeSigma™ Supelco™ Sephadex™ G-100, Medium
Sephadex™ G-100 is a gel filtration medium used in protein chromatography, affinity chromatography and gel filtration chromatography.