CAS RN 90530-31-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Octyl Acrylate (mixture of branched chain isomers) (stabilized with MEHQ), TCI America™
CAS: 90530-31-7 Molecular Formula: C11H20O2 Synonym: Acrylic Acid Octyl Ester