CAS RN 9056-59-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Amberlite IRA-68, ion exchange resin, Thermo Scientific™
CAS: 9056-59-1 MDL Number: MFCD00145795