accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90561-85-6

IUPAC Name: 2-bromo-1,3,5-trimethyl-4-nitrobenzene
Synonyms: 2-bromo-1,3,5-trimethyl-4-nitrobenzene
Molecular Weight (g/mol): 244.09
Molecular Formula: C9H10BrNO2
InChi Key: IUFDRRDWXUSFBG-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(Br)C(C)=C1[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results