accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90564-77-5

IUPAC Name: 1-(3-phenyl-1,2,4-oxadiazol-5-yl)methanamine
Synonyms: 1-(3-phenyl-1,2,4-oxadiazol-5-yl)methanamine
Molecular Weight (g/mol): 175.19
Molecular Formula: C9H9N3O
InChi Key: QFBMJBDECSEYCZ-UHFFFAOYSA-N
SMILES: NCC1=NC(=NO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results