accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOHNClFFFCl

CAS RN 905711-97-9

IUPAC Name: N-[2,6-dichloro-4-(trifluoromethyl)phenyl]benzenesulfonamide
Synonyms: N-[2,6-dichloro-4-(trifluoromethyl)phenyl]benzenesulfonamide
Molecular Weight (g/mol): 370.17
Molecular Formula: C13H8Cl2F3NO2S
InChi Key: DHBRZEHYRUKIHH-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(NS(=O)(=O)C2=CC=CC=C2)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results