accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOH2NOOH

CAS RN 905954-28-1

IUPAC Name: 1-amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid
Synonyms: 1-amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid
Molecular Weight (g/mol): 353.41
Molecular Formula: C15H31NO8
InChi Key: PVRGRRPCHFTMMD-UHFFFAOYSA-N
SMILES: NCCOCCOCCOCCOCCOCCOCCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)