CAS RN 906352-85-0
IUPAC Name:
3-(1-methyl-1H-pyrazol-3-yl)benzoic acid
Synonyms:
3-(1-methylpyrazol-3-yl)benzoic acid
Molecular Weight (g/mol):
202.21
Molecular Formula:
C11H10N2O2
InChi Key:
NQPMBEUYTBLTAG-UHFFFAOYSA-N
SMILES:
CN1C=CC(=N1)C1=CC(=CC=C1)C(O)=O
1
–
1
of
1
results
1
3-(1-Methyl-1H-pyrazol-3-yl)benzoic acid, 97%, Thermo Scientific™
CAS: 906352-85-0 Molecular Formula: C11H10N2O2 Molecular Weight (g/mol): 202.213 MDL Number: MFCD09065006 InChI Key: NQPMBEUYTBLTAG-UHFFFAOYSA-N Synonym: 3-1-methyl-1h-pyrazol-3-yl benzoic acid,3-1-methylpyrazol-3-yl benzoic acid,benzoicacid,3-1-methyl-1h-pyrazol-3-yl,3-3-carboxyphenyl-1-methyl-1h-pyrazole,benzoicacid, 3-1-methyl-1h-pyrazol-3-yl,benzoic acid, 3-1-methyl-1h-pyrazol-3-yl PubChem CID: 24229627 IUPAC Name: 3-(1-methylpyrazol-3-yl)benzoic acid SMILES: CN1C=CC(=N1)C2=CC(=CC=C2)C(=O)O