accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOOOH

CAS RN 906353-00-2

IUPAC Name: 4-[(6-methylpyrazin-2-yl)oxy]benzoic acid
Synonyms: 4-[(6-methylpyrazin-2-yl)oxy]benzoic acid
Molecular Weight (g/mol): 230.22
Molecular Formula: C12H10N2O3
InChi Key: PPGBPVGCYRDKMI-UHFFFAOYSA-N
SMILES: CC1=CN=CC(OC2=CC=C(C=C2)C(O)=O)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results