accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOO

CAS RN 906353-01-3

IUPAC Name: 4-[(6-methylpyrazin-2-yl)oxy]benzaldehyde
Synonyms: 4-[(6-methylpyrazin-2-yl)oxy]benzaldehyde
Molecular Weight (g/mol): 214.22
Molecular Formula: C12H10N2O2
InChi Key: VUPYANPNFUPQQU-UHFFFAOYSA-N
SMILES: CC1=CN=CC(OC2=CC=C(C=O)C=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results