accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 906422-74-0

IUPAC Name: 1-[4-(thiophen-2-yl)oxan-4-yl]methanamine
Synonyms: 1-[4-(thiophen-2-yl)oxan-4-yl]methanamine
Molecular Weight (g/mol): 197.3
Molecular Formula: C10H15NOS
InChi Key: LBEBHUWSRFXHQR-UHFFFAOYSA-N
SMILES: NCC1(CCOCC1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results