accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 906478-91-9

IUPAC Name: triethyl(pentyl)azanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: bistriflylimide anion; triethyl(pentyl)azanium
Molecular Weight (g/mol): 452.47
Molecular Formula: C13H26F6N2O4S2
InChi Key: ALYCOCULEAWWJO-UHFFFAOYSA-N
SMILES: CCCCC[N+](CC)(CC)CC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results