accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HO

CAS RN 90676-25-8

IUPAC Name: 2-propylcyclohexan-1-ol
Synonyms: 2-propylcyclohexan-1-ol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: VZBNUCDUQJCIDP-UHFFFAOYNA-N
SMILES: CCCC1CCCCC1O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results