accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90729-43-4

IUPAC Name: 1-(4-tert-butylphenyl)-4-[4-(diphenylmethoxy)piperidin-1-yl]butan-1-one
Synonyms: kestin pharmafarm brand of ebastine bactil cepa brand of ebastine almirall brand of ebastine ebastinum busidril ebastel bayer brand of ebastine evastel
Molecular Weight (g/mol): 469.67
Molecular Formula: C32H39NO2
InChi Key: MJJALKDDGIKVBE-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C(=O)CCCN1CCC(CC1)OC(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)