CAS RN 9073-60-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ β-Lactamase, Bacillus cereus 569/H9
Useful for assaying penicillins and cephalosporins