CAS RN 908294-68-8
CAS RN 908294-68-8
IUPAC Name:
N-[2,6-bis(propan-2-yl)phenyl]-1-(pyridin-2-yl)methanimine
Synonyms:
N-(2,6-diisopropylphenyl)-1-pyridin-2-ylmethanimine
Molecular Weight (g/mol):
266.39
Molecular Formula:
C18H22N2
InChi Key:
BABQFMPURPKJNW-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=CC(C(C)C)=C1N=CC1=CC=CC=N1
1
–
1
of
1
results
1
trans-2,6-Diisopropyl-N-(2-pyridylmethylene)aniline 95.0+%, TCI America™

CAS: 908294-68-8 Molecular Formula: C18H22N2 Molecular Weight (g/mol): 266.39 MDL Number: MFCD06669887 InChI Key: BABQFMPURPKJNW-UHFFFAOYSA-N PubChem CID: 1284716 IUPAC Name: N-[2,6-bis(propan-2-yl)phenyl]-1-(pyridin-2-yl)methanimine SMILES: CC(C)C1=CC=CC(C(C)C)=C1N=CC1=CC=CC=N1