accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90843-31-5

IUPAC Name: 1-(2,3-dihydro-1-benzofuran-5-yl)ethan-1-one
Synonyms: 1-(2,3-dihydro-1-benzofuran-5-yl)ethanone
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: MMVUJVASBDVNGJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2OCCC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results