accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNSH3C

CAS RN 90921-53-2

IUPAC Name: 2,2-dimethyl-3,4-dihydro-2H-1,4-benzothiazin-3-one
Synonyms: 2,2-dimethyl-4H-1,4-benzothiazin-3-one
Molecular Weight (g/mol): 193.26
Molecular Formula: C10H11NOS
InChi Key: KQXBYDQIOLXEDZ-UHFFFAOYSA-N
SMILES: CC1(C)SC2=CC=CC=C2NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results