accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 909280-97-3

IUPAC Name: 5,11-dibromo-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Synonyms: 5,11-dibromo-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Molecular Weight (g/mol): 348.07
Molecular Formula: C10H4Br2S2
InChi Key: SXRSYJAHJIIXFM-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC3=C(C=C(Br)S3)C=C2S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results