accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90965-06-3

IUPAC Name: 1-(diazyn-1-ium-1-yl)-1-(dimethoxyphosphoryl)prop-1-en-2-olate
Synonyms: seyferth-gilbert homologation
Molecular Weight (g/mol): 192.11
Molecular Formula: C5H9N2O4P
InChi Key: SQHSJJGGWYIFCD-UHFFFAOYSA-N
SMILES: COP(=O)(OC)C([N+]#N)=C(C)[O-]
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)