accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91-01-0

IUPAC Name: diphenylmethanol
Synonyms: benzhydrol
Molecular Weight (g/mol): 184.24
Molecular Formula: C13H12O
InChi Key: QILSFLSDHQAZET-UHFFFAOYSA-N
SMILES: OC(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (8)
Boiling Point 
  • (3)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (2)
  • (2)
  • (8)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)