accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91-09-8

IUPAC Name: (2S,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
Synonyms: (2S,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
Molecular Weight (g/mol): 164.16
Molecular Formula: C6H12O5
InChi Key: ZBDGHWFPLXXWRD-MOJAZDJTSA-N
SMILES: CO[C@H]1OC[C@@H](O)[C@H](O)[C@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results