accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 91-19-0

IUPAC Name: quinoxaline
Synonyms: quinoxaline quinazine benzopyrazine benzoparadiazine benzo[a]pyrazine benzo(a)pyrazine phenpiazine phenopiazine chinoxalin
Molecular Weight (g/mol): 130.15
Molecular Formula: C8H6N2
InChi Key: XSCHRSMBECNVNS-UHFFFAOYSA-N
SMILES: C1=CC=C2N=CC=NC2=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)