accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO(E)

CAS RN 91-48-5

IUPAC Name: (2E)-2,3-diphenylprop-2-enoate
Synonyms: α-phenylcinnamic acid
Molecular Weight (g/mol): 223.25
Molecular Formula: C15H11O2
InChi Key: BIDDLDNGQCUOJQ-SDNWHVSQSA-M
SMILES: [O-]C(=O)C(=C\C1=CC=CC=C1)\C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results