accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91-56-5

IUPAC Name: 2,3-dihydro-1H-indole-2,3-dione
Synonyms: isatin isotin 2,3-ketoindoline o-aminobenzoylformic anhydride pseudoisatin isatic acid lactam tribulin
Molecular Weight (g/mol): 147.13
Molecular Formula: C8H5NO2
InChi Key: JXDYKVIHCLTXOP-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (5)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (5)
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (5)
  • (5)
  • (6)
  • (3)
  • (3)
  • (5)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)