accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91-65-6

IUPAC Name: N,N-diethylcyclohexanamine
Synonyms: cyclohexanamine, N,N-diethyl-
Molecular Weight (g/mol): 155.29
Molecular Formula: C10H21N
InChi Key: CIXSDMKDSYXUMJ-UHFFFAOYSA-N
SMILES: CCN(CC)C1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results