accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91-66-7

IUPAC Name: N,N-diethylaniline
Synonyms: diethylaniline
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: GGSUCNLOZRCGPQ-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (8)
Percent Purity 
  • (3)
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (8)
  • (3)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)