accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNN

CAS RN 91-78-1

IUPAC Name: 1,3,5-triphenyl-1,3,5-triazinane
Synonyms: 1,3,5-triphenyl-1,3,5-triazinane
Molecular Weight (g/mol): 315.42
Molecular Formula: C21H21N3
InChi Key: VASMRQAVWVVDPA-UHFFFAOYSA-N
SMILES: C1N(CN(CN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results