accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCONCOCH3

CAS RN 91-97-4

IUPAC Name: 4,4'-diisocyanato-3,3'-dimethyl-1,1'-biphenyl
Synonyms: 4,4'-diisocyanato-3,3'-dimethyl-1,1'-biphenyl
Molecular Weight (g/mol): 264.28
Molecular Formula: C16H12N2O2
InChi Key: ICLCCFKUSALICQ-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1N=C=O)C1=CC=C(N=C=O)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results