accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNHNOOOHHOOOOHHO

CAS RN 910036-85-0

IUPAC Name: [(1H-1,3-benzodiazol-6-yl)methyl](methyl)amine; bis(oxalic acid)
Synonyms: (3H-1,3-benzodiazol-5-ylmethyl)(methyl)amine; bis(oxalic acid)
Molecular Weight (g/mol): 341.28
Molecular Formula: C13H15N3O8
InChi Key: HDQRBNFLWRHBAA-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.OC(=O)C(O)=O.CNCC1=CC=C2N=CNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results