accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 910036-89-4

NFFFOOFFFFF

CAS RN 910036-89-4

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 4-[5-(trifluoromethyl)pyridin-2-yl]benzoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 4-[5-(trifluoromethyl)pyridin-2-yl]benzoate
Molecular Weight (g/mol): 433.26
Molecular Formula: C19H7F8NO2
InChi Key: FXVFNQGTIWSDPI-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CC=C(C=C2)C2=NC=C(C=C2)C(F)(F)F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results