CAS RN 910036-94-1
IUPAC Name:
tert-butyl N-[(2-bromothiophen-3-yl)methyl]carbamate
Synonyms:
tert-butyl N-[(2-bromothiophen-3-yl)methyl]carbamate
Molecular Weight (g/mol):
292.19
Molecular Formula:
C10H14BrNO2S
InChi Key:
RFNWODFBZMWIEH-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NCC1=C(Br)SC=C1
1
–
1
of
1
results
1
tert-Butyl (2-bromothien-3-yl)methylcarbamate, 95%, Thermo Scientific™
CAS: 910036-94-1 Molecular Formula: C10H14BrNO2S Molecular Weight (g/mol): 292.191 MDL Number: MFCD09025882 InChI Key: RFNWODFBZMWIEH-UHFFFAOYSA-N Synonym: tert-butyl 2-bromothien-3-yl methylcarbamate,tert-butyl n-2-bromothiophen-3-yl methyl carbamate,tert-butyl 2-bromothiophen-3-yl methyl carbamate,tert-butyl 2-bromothiophen-3-yl methylcarbamate PubChem CID: 24229570 IUPAC Name: tert-butyl N-[(2-bromothiophen-3-yl)methyl]carbamate SMILES: CC(C)(C)OC(=O)NCC1=C(SC=C1)Br