accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONOOFFFFF

CAS RN 910036-96-3

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 6-(oxan-4-yloxy)pyridine-3-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl 6-(oxan-4-yloxy)pyridine-3-carboxylate
Molecular Weight (g/mol): 389.28
Molecular Formula: C17H12F5NO4
InChi Key: DOADFHHFWSVLLM-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CN=C(OC3CCOCC3)C=C2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results