accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONCO

CAS RN 910037-00-2

IUPAC Name: 1-isocyanato-2-(phenoxymethyl)benzene
Synonyms: 1-isocyanato-2-(phenoxymethyl)benzene
Molecular Weight (g/mol): 225.25
Molecular Formula: C14H11NO2
InChi Key: FGEXXDXHIUWRRS-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=CC=C1COC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results