accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 910037-02-4

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 3-(1,3-dioxolan-2-yl)thiophene-2-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl 3-(1,3-dioxolan-2-yl)thiophene-2-carboxylate
Molecular Weight (g/mol): 366.26
Molecular Formula: C14H7F5O4S
InChi Key: FHURIHOENRMCSL-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=C(C=CS2)C2OCCO2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results