accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 910037-11-5

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 3-(1-methyl-1H-pyrazol-3-yl)benzoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 3-(1-methylpyrazol-3-yl)benzoate
Molecular Weight (g/mol): 368.26
Molecular Formula: C17H9F5N2O2
InChi Key: LGHVUZDAZTZCHC-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C1=CC(=CC=C1)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results