accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONBr

CAS RN 910037-14-8

IUPAC Name: 7-bromo-4-methyl-2H,3H,4H-pyrido[3,2-b][1,4]oxazine
Synonyms: 7-bromo-4-methyl-2H,3H-pyrido[3,2-b][1,4]oxazine
Molecular Weight (g/mol): 229.08
Molecular Formula: C8H9BrN2O
InChi Key: KPGNYZHIKPSZMS-UHFFFAOYSA-N
SMILES: CN1CCOC2=CC(Br)=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results